Läs mer
7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone, TRC
Produktkod. 30136136
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136136 1 g
17951475 50 mg
17961455 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136136 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3655601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120004-79-7
Molekylformel C13 H16 Cl N O2
Molekylvikt (g/mol) 253.72
Synonym 2(1H)-Quinolinone, 7-(4-chlorobutoxy)-3,4-dihydro-, 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone, 7-(4-Chlorobutoxy)-3,4-dihydrocarbostyril
IUPAC-namn 7-(4-chlorobutoxy)-3,4-dihydro-1H-quinolin-2-one
LEDER ClCCCCOc1ccc2CCC(=O)Nc2c1
Specifikationer
Kemiskt namn eller material 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone
Kvantitet 1 g
InChI formel InChI=1S/C13H16ClNO2/c14-7-1-2-8-17-11-5-3-10-4-6-13(16)15-12(10)9-11/h3,5,9H,1-2,4,6-8H2,(H,15,16)
Formel vikt 253.087