Läs mer
7-(4-Chlorobutoxy)quinolin-2(1H)-one, TRC
Produktkod. 30352554
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30352554 2.5 g 2.5g
30366235 250 mg -
30352555 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352554 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3813432.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 913613-82-8
Molekylformel C13H14ClNO2
Molekylvikt (g/mol) 251.71
Synonym 7-(4-Chlorobutoxy)-2(1H)-quinolinone, 7-(4-Chlorobutoxy)-1H-quinolin-2-one
IUPAC-namn 7-(4-chlorobutoxy)-1H-quinolin-2-one
LEDER ClCCCCOc1ccc2C=CC(=O)Nc2c1
Specifikationer
Kemiskt namn eller material 7-(4-Chlorobutoxy)quinolin-2(1H)-one
Kvantitet 2.5 g
InChI formel InChI=1S/C13H14ClNO2/c14-7-1-2-8-17-11-5-3-10-4-6-13(16)15-12(10)9-11/h3-6,9H,1-2,7-8H2,(H,15,16)
Formel vikt 251.0713