Läs mer
7,8,9,10-Tetrahydro-6,10-Methano-6H-pyrazino[2,3-h][3]benzazepine 1,4-Dioxide, TRC
Produktkod. 30422771
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422771 50 mg
30420793 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422771 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29395550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1337563-47-9
Molekylformel C13H13N3O2
Molekylvikt (g/mol) 243.26
LEDER [O-][n+]1cc[n+]([O-])c2cc3C4CNCC(C4)c3cc12
Specifikationer
Kemiskt namn eller material 7,8,9,10-Tetrahydro-6,10-Methano-6H-pyrazino[2,3-h][3]benzazepine 1,4-Dioxide
Kvantitet 50 mg
InChI formel InChI=1S/C13H13N3O2/c17-15-1-2-16(18)13-5-11-9-3-8(6-14-7-9)10(11)4-12(13)15/h1-2,4-5,8-9,14H,3,6-7H2
Formel vikt 243.1008