Läs mer
7,8-Dihydroxy-2,3,4,5-tetrahydro-2-benzazepine, Hydrobromide, TRC
Produktkod. 30375445
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30375445 25 mg
1 options
Produktkod. 30375445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45502525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7,8-Dihydroxy-2,3,4,5-tetrahydro-2-benzazepine, Hydrobromide
CAS 113853-92-2
Molekylformel C10 H13 N O2 . Br H
Kvantitet 25 mg
Synonym 1H-2-Benzazepine-7,8-diol, 2,3,4,5-tetrahydro-, hydrobromide (1:1), 1H-2-Benzazepine-7,8-diol, 2,3,4,5-tetrahydro-, hydrobromide (9CI), 2,3,4,5-Tetrahydro-1H-benzo[c]azepine-7,8-diol hydrobromide, 2,3,4,5-Tetrahydro-7,8-dihydroxy-1H-2-benzazepine hydrobromide, 7,8-Dihydroxy-2,3,4,5-tetrahydro-2-benzazepine hydrobromide
InChI formel InChI=1S/C10H13NO2.BrH/c12-9-4-7-2-1-3-11-6-8(7)5-10(9)13;/h4-5,11-13H,1-3,6H2;1H
LEDER Br.Oc1cc2CCCNCc2cc1O
IUPAC-namn 2,3,4,5-tetrahydro-1H-2-benzazepine-7,8-diol;hydrobromide
Molekylvikt (g/mol) 260.13
Formel vikt 259.0208