Läs mer
7,9-Dimethylguanine (>80%), TRC
Produktkod. 30240893
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240893 10 mg
30240523 5 mg
30240676 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240893 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47312810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 524-35-6
Molekylformel C7H9N5O
Molekylvikt (g/mol) 179.18
LEDER C[n+]1cn(C)c2c([O-])nc(N)nc12
Specifikationer
Kemiskt namn eller material 7,9-Dimethylguanine