Läs mer
7,9-Dimethylguanine Hemitosylate, TRC
Produktkod. 30378402
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30378402 100 mg
1 options
Produktkod. 30378402 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD473125100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7,9-Dimethylguanine Hemitosylate
CAS 12240-99-2
Molekylformel 2 C7 H9 N5 O . C7 H8 O3 S
Kvantitet 100 mg
Synonym 2-Amino-6-hydroxy-7,9-dimethyl-purinium p-Toluenesulfonate, Herbipoline Hemitosylate, 2-Amino-1,6-dihydro-7,9-dimethyl-6-oxopurinium Hydroxide Hemitosylate
InChI formel InChI=1S/2C7H9N5O.C7H8O3S/c2*1-11-3-12(2)5-4(11)6(13)10-7(8)9-5;1-6-2-4-7(5-3-6)11(8,9)10/h2*3H,1-2H3,(H2-,8,9,10,13);2-5H,1H3,(H,8,9,10)
LEDER Cc1ccc(cc1)S(=O)(=O)O.C[n+]2cn(C)c3nc(N)nc([O-])c23.C[n+]4cn(C)c5nc(N)nc([O-])c45
IUPAC-namn 2-amino-7,9-dimethylpurin-7-ium-6-olate;4-methylbenzenesulfonic acid
Molekylvikt (g/mol) 530.56
Formel vikt 530.181