missing translation for 'onlineSavingsMsg'
Läs mer

7-Acetyl-5-oxo-5H-[1]benzopyrano[2,3-b]pyridine, TRC

Produktkod. 30309430
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30309430 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30309430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18787025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 7-Acetyl-5-oxo-5H-[1]benzopyrano[2,3-b]pyridine
CAS 53944-40-4
Molekylformel C14H9NO3
Kvantitet 25 mg
Synonym 7-Acetyl-5H-[1]Benzopyrano[2,3-b]pyridin-5-one
InChI formel InChI=1S/C14H9NO3/c1-8(16)9-4-5-12-11(7-9)13(17)10-3-2-6-15-14(10)18-12/h2-7H,1H3
LEDER CC(=O)c1ccc2Oc3ncccc3C(=O)c2c1
IUPAC-namn 7-acetylchromeno[2,3-b]pyridin-5-one
Molekylvikt (g/mol) 239.23
Formel vikt 239.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.