Läs mer
7-Amino-1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid, TRC
Produktkod. 30006320
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30006320 2.5 g 2.5g
17902795 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006320 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6037802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 172426-88-9
Molekylformel C14 H13 F N2 O4
Molekylvikt (g/mol) 292.26
Synonym 3-Quinolinecarboxylic acid, 7-amino-1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-4-oxo-, 7-Amino-1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic acid
IUPAC-namn 7-amino-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid
LEDER COc1c(N)c(F)cc2C(=O)C(=CN(C3CC3)c12)C(=O)O
Specifikationer
Kemiskt namn eller material 7-Amino-1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid
Analyt- eller komponentnamn 7-Amino-1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C14H13FN2O4/c1-21-13-10(16)9(15)4-7-11(13)17(6-2-3-6)5-8(12(7)18)14(19)20/h4-6H,2-3,16H2,1H3,(H,19,20)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 2.5 g
Formel vikt 292.0859
Procent renhet >95%