missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-3-vinyl-3-cephem-4-carboxylic Acid, TRC

Product Code. 30334514
Change view
Click to view available options
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30334514 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334514 Supplier Toronto Research Chemicals Supplier No. TRCA6326005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 7-Amino-3-vinyl-3-cephem-4-carboxylic Acid
CAS 79349-82-9
Molekylformel C9 H10 N2 O3 S
Kvantitet 5 g
Synonym (6R,7R)-7-Amino-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, (6R-trans)-7-Amino-3-ethenyl-8-oxo-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, 7-AVCA
InChI formel InChI=1S/C9H10N2O3S/c1-2-4-3-15-8-5(10)7(12)11(8)6(4)9(13)14/h2,5,8H,1,3,10H2,(H,13,14)/t5-,8-/m1/s1
LEDER N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)C=C
IUPAC-namn (6R,7R)-7-amino-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molekylvikt (g/mol) 226.25
Formel vikt 226.0412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.