missing translation for 'onlineSavingsMsg'
Läs mer

7-Aminobenzo[a]pyrene, TRC

Produktkod. 30315979
Change view
Click to view available options
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30315979 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315979 Supplier Toronto Research Chemicals Supplier No. TRCA5928501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 7-Aminobenzo[a]pyrene
CAS 72297-05-3
Molekylformel C20 H13 N
Kvantitet 1 mg
Synonym Benzo[a]pyren-7-amine, 7-Aminobenzo[a]pyrene
InChI formel InChI=1S/C20H13N/c21-18-6-2-5-15-16-10-9-13-4-1-3-12-7-8-14(11-17(15)18)20(16)19(12)13/h1-11H,21H2
LEDER Nc1cccc2c1cc3ccc4cccc5ccc2c3c45
IUPAC-namn benzo[a]pyren-7-amine
Molekylvikt (g/mol) 267.32
Formel vikt 267.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.