missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
7-Aminodesacetoxycephalosporanic acid, 98%, Thermo Scientific™
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
7-Aminodesacetoxycephalosporanic acid is the Cephalexin metabolite. It is a Potent inhibitor of bacterial cell-wall enzyme. It is used in the synthesis of cephalosporins and for bioconversion studies.
Solubility
Soluble in water (partly), 1M ammonium hydroxide (50 mg/ml), and DMSO (Sparingly).
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
7-Aminodesacetoxycephalosporanic acid is the Cephalexin metabolite. It is a Potent inhibitor of bacterial cell-wall enzyme. It is used in the synthesis of cephalosporins and for bioconversion studies.
Solubility
Soluble in water (partly), 1M ammonium hydroxide (50 mg/ml), and DMSO (Sparingly).
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 7-Aminodesacetoxycephalosporanic acid |
| Smältpunkt | ∽234°C (decomposition) |
| Kvantitet | 5 g |
| Beilstein | 5273616 |
| Löslighetsinformation | Soluble in water (partly),1M ammonium hydroxide (50mg/ml),and DMSO (Sparingly). |
| Formel vikt | 214.24 |
| Procent renhet | 98% |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?