Läs mer
7-Aminoheptanoic Acid Hydrochloride, TRC
Produktkod. 30328607
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30328607 50 g
1 options
Produktkod. 30328607 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60991050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-Aminoheptanoic Acid Hydrochloride
CAS 62643-56-5
Molekylformel C7H16ClNO2
Kvantitet 50 g
Synonym 7-Aminoenanthic Acid Hydrochloride, NSC 59008 Hydrochloride, zeta-Aminoenanthic Acid Hydrochloride, ω-Aminoenanthic Acid Hydrochloride, ω-Aminoheptanoic Acid Hydrochloride
InChI formel InChI=1S/C7H15NO2.ClH/c8-6-4-2-1-3-5-7(9)10;/h1-6,8H2,(H,9,10);1H
LEDER Cl.NCCCCCCC(=O)O
IUPAC-namn 7-aminoheptanoic acid;hydrochloride
Molekylvikt (g/mol) 181.66
Formel vikt 181.087