missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7033039
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
754214-56-7
C13H17BN2O2
244.10
MFCD08060937
UOXAMYZTYZLSCC-UHFFFAOYSA-N
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p
24208789
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1
This item is not returnable. View return policy
754214-56-7
C13H17BN2O2
244.10
MFCD08060937
UOXAMYZTYZLSCC-UHFFFAOYSA-N
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p
24208789
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1

7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, ethanol and THF.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 241°C to 245°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Soluble in methanol,ethanol and THF.
Formula Weight 244.1
Percent Purity 97%
Chemical Name or Material 7-Azaindole-5-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'