missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoquinoline, 95%, Thermo Scientific Chemicals

Catalog Number 15450989
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
4965-36-0
C9H6BrN
208.058
MFCD03695823
XYBSZCUHOLWQQU-UHFFFAOYSA-N
quinoline, 7-bromo, 7-bromo-quinoline, pubchem12788, ksc238o9n, abbypharma ap-12-10883
521259
7-bromoquinoline
C1=CC2=C(C=C(C=C2)Br)N=C1
This item is not returnable. View return policy
4965-36-0
C9H6BrN
208.058
MFCD03695823
XYBSZCUHOLWQQU-UHFFFAOYSA-N
quinoline, 7-bromo, 7-bromo-quinoline, pubchem12788, ksc238o9n, abbypharma ap-12-10883
521259
7-bromoquinoline
C1=CC2=C(C=C(C=C2)Br)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 208.06
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Bromoquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'