Läs mer
7-Carboxy-5-chloro-8-hydroxy-3,4-dihydro-3-methylisocoumarin, TRC
Produktkod. 17961625
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17961625 10 mg
17911325 1 mg
17924145 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17961625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17786510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16281-39-3
Molekylformel C11 H9 Cl O5
Molekylvikt (g/mol) 256.639
Synonym 3-Methyl-5-chloro-8-hydroxy-3,4-dihydroisocoumarin-7-carboxylic Acid, 5-Chloro-3,4-dihydro-8-hydroxy-3-methylisocoumarin-7-carboxylic Acid, 5-Chloro-8-hydroxy-3,4-dihydro-3-methylisocoumarin-7-carboxylic Acid, 5-Chloro-8-hydroxy-3-methyl-1-oxo-7-isochromancarboxylic Acid, 5-Chloro-3,4-dihydro-8-hydroxy-3-methyl-1-oxo-1H-2-benzopyran-7-carboxylic Acid
IUPAC-namn 5-chloro-8-hydroxy-3-methyl-1-oxo-isochromane-7-carboxylic acid
LEDER CC1Cc2c(Cl)cc(C(=O)O)c(O)c2C(=O)O1
Specifikationer
Kemiskt namn eller material 7-Carboxy-5-chloro-8-hydroxy-3,4-dihydro-3-methylisocoumarin
Kvantitet 10 mg
InChI formel InChI=1S/C11H9ClO5/c1-4-2-5-7(12)3-6(10(14)15)9(13)8(5)11(16)17-4/h3-4,13H,2H2,1H3,(H,14,15)
Formel vikt 256.0139