missing translation for 'onlineSavingsMsg'
Läs mer

7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one, TRC

Produktkod. 30365165
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365165 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30365165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36626525G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one
CAS 160129-45-3
Molekylformel C10H10ClNO
Kvantitet 25 g
Synonym 7-Chloro-5-oxo-2,3,4,5-tetrahydro-1H-1-benzazepine
InChI formel InChI=1S/C10H10ClNO/c11-7-3-4-9-8(6-7)10(13)2-1-5-12-9/h3-4,6,12H,1-2,5H2
LEDER Clc1ccc2NCCCC(=O)c2c1
IUPAC-namn 7-chloro-1,2,3,4-tetrahydro-1-benzazepin-5-one
Molekylvikt (g/mol) 195.65
Formel vikt 195.0451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.