Läs mer
7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride, TRC
Produktkod. 30352881
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30352881 25 mg
1 options
Produktkod. 30352881 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36603025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride
CAS 526204-10-4
Molekylformel C17 H18 Cl N3 O3 . Cl H
Kvantitet 25 mg
Synonym 7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride, Ciprofloxacin Related Compound A (USP), 3-Quinolinecarboxylic acid, 7-chloro-1-cyclopropyl-1,4-dihydro-4-oxo-6-(1-piperazinyl)-, monohydrochloride (9CI)
InChI formel InChI=1S/C17H18ClN3O3.ClH/c18-13-8-14-11(7-15(13)20-5-3-19-4-6-20)16(22)12(17(23)24)9-21(14)10-1-2-10;/h7-10,19H,1-6H2,(H,23,24);1H
LEDER Cl.OC(=O)C1=CN(C2CC2)c3cc(Cl)c(cc3C1=O)N4CCNCC4
IUPAC-namn 7-chloro-1-cyclopropyl-4-oxo-6-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 384.26
Formel vikt 383.0803