missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Mikromol™

Product Code. 30052601
Change view
Click to view available options
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30052601 25 mg
17948334 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052601 Supplier Mikromol Supplier No. MM0350.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS 68077-26-9
Analyt- eller komponentnamn 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Frakt skick Room Temperature
Molekylformel C12 H9 Cl F N O3
Certifikat och efterlevnad ISO 17025
Synonym 7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Norfloxacin Imp. A (EP), Norfloxacin USP Related Compound A, Norfloxacin USP RC A, Norfloxacin Related Compound A, Norfloxacin Impurity A, Pefloxacin Mesilate Dihydrate Impurity F, Pefloxacin Impurity F
InChI formel InChI=1S/C12H9ClFNO3/c1-2-15-5-7(12(17)18)11(16)6-3-9(14)8(13)4-10(6)15/h3-5H,2H2,1H3,(H,17,18)
LEDER CCN1C=C(C(=O)O)C(=O)c2cc(F)c(Cl)cc12
IUPAC-namn 7-chloro-1-ethyl-6-fluoro-4-oxoquinoline-3-carboxylic acid
Molekylvikt (g/mol) 269.66
Kvantitet 25 mg
Formel vikt 269.0255 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.