Läs mer
7-Chloro-5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide (Demoxepam), Mikromol™
Produktkod. 17934266
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17934266 100 mg
17952936 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17934266 Leverantör Mikromol Leverantörsnummer MM0028.02
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-Chloro-5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide (Demoxepam)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Demoxepam
CAS 963-39-3
Analyt- eller komponentnamn Demoxepam
Frakt skick Room Temperature
Molekylformel C15 H11 Cl N2 O2
Certifikat och efterlevnad ISO 17025
Synonym Chlordiazepoxide Hydrochloride Imp. A (EP), Chlordiazepoxide Imp. A (EP), Oxazepam Imp. E (EP), Chlordiazepoxide USP Related Compound A, Chlordiazepoxide USP RC A, 7-Chloro-5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide, Demoxepam, Chlordiazepoxide Related Compound A, Chlordiazepoxide Hydrochloride Impurity A, Chlordiazepoxide Impurity A, Oxazepam Impurity E, Chlordiazepoxide Impurity A
InChI formel InChI=1S/C15H11ClN2O2/c16-11-6-7-13-12(8-11)15(10-4-2-1-3-5-10)18(20)9-14(19)17-13/h1-8H,9H2,(H,17,19)
LEDER [O-][N+]1=C(c2ccccc2)c3cc(Cl)ccc3NC(=O)C1
IUPAC-namn 7-chloro-4-oxido-5-phenyl-1,3-dihydro-1,4-benzodiazepin-4-ium-2-one
Molekylvikt (g/mol) 286.71
Kvantitet 100 mg
Formel vikt 286.0509 g/mol