Läs mer
7-Chloro-imidazo[1,2-C]pyrimidin-5-ol, TRC
Produktkod. 30352234
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352234 100 mg
30360417 10 mg
30361010 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352234 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC384658100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56817-09-5
Molekylformel C6H4N3OCl
Molekylvikt (g/mol) 169.56
Synonym Imidazo[1,2-c]pyrimidin-5(6H)-one, 7-chloro- (9CI, ACI), 7-Chloroimidazo[1,2-c]pyrimidin-5(6H)-one (ACI), 7-Chloro-imidazo[1,2-c]pyrimidin-5-ol
IUPAC-namn 7-chloro-6H-imidazo[1,2-c]pyrimidin-5-one
LEDER ClC1=Cc2nccn2C(=O)N1
Specifikationer
Kemiskt namn eller material 7-Chloro-imidazo[1,2-C]pyrimidin-5-ol
Kvantitet 100 mg
InChI formel InChI=1S/C6H4ClN3O/c7-4-3-5-8-1-2-10(5)6(11)9-4/h1-3H,(H,9,11)
Formel vikt 169.0043