missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroquinoline-4-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number p-7033859
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
871125-83-6
C15H17BClNO2
289.57
MFCD07783020
GZDUYRAAWRRVEY-UHFFFAOYSA-N
7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloroquinoline-4-boronic acid pinacol ester, 7-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline,7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 7-chloroquinoline-4-boronic acid, pinacol ester, 7-chloroquinolin-4-yl boronic acid pinacol ester
16218076
7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
CC1(C)OB(OC1(C)C)C1=CC=NC2=CC(Cl)=CC=C12
This item is not returnable. View return policy
871125-83-6
C15H17BClNO2
289.57
MFCD07783020
GZDUYRAAWRRVEY-UHFFFAOYSA-N
7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloroquinoline-4-boronic acid pinacol ester, 7-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline,7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 7-chloroquinoline-4-boronic acid, pinacol ester, 7-chloroquinolin-4-yl boronic acid pinacol ester
16218076
7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
CC1(C)OB(OC1(C)C)C1=CC=NC2=CC(Cl)=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 289.56
Percent Purity 96%
Chemical Name or Material 7-Chloroquinoline-4-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'