Läs mer
7-Cyanomethotrexate Dimethyl Ester, TRC
Produktkod. 30367304
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30367304 1 mg
1 options
Produktkod. 30367304 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9818201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-Cyanomethotrexate Dimethyl Ester
CAS 112163-39-0
Molekylformel C23 H25 N9 O5
Kvantitet 1 mg
Synonym L-Glutamic acid, N-[4-[[(2,4-diamino-7-cyano-6-pteridinyl)methyl]methylamino]benzoyl]-, dimethyl ester (9CI), (S)-Dimethyl 2-(4-(((4-amino-7-cyano-2-imino-2,3-dihydropteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioate, (2S)-2-[[4-[[(2,4-Diamino-7-cyanopteridin-6-yl)methyl]methylamino]benzoyl]amino]pentanedioic acid dimethyl ester, N-[4-[[(2,4-Diamino-7-cyano-7,8-dihydro-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic acid dimethyl ester, 7-Cyanomethotrexate dimethyl ester
InChI formel InChI=1S/C23H25N9O5/c1-32(11-16-15(10-24)28-20-18(27-16)19(25)30-23(26)31-20)13-6-4-12(5-7-13)21(34)29-14(22(35)37-3)8-9-17(33)36-2/h4-7,14H,8-9,11H2,1-3H3,(H,29,34)(H4,25,26,28,30,31)/t14-/m0/s1
LEDER COC(=O)CC[C@H](NC(=O)c1ccc(cc1)N(C)Cc2nc3c(N)nc(N)nc3nc2C#N)C(=O)OC
IUPAC-namn dimethyl (2S)-2-[[4-[(2,4-diamino-7-cyanopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioate
Molekylvikt (g/mol) 507.5
Formel vikt 507.1979