missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Deazahypoxanthine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Moisture and light sensitive. Store at -20°C
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Moisture and light sensitive. Store at -20°C
Specifications
Melting Point | 345°C to 350°C |
Color | Brown |
Quantity | 1 g |
Beilstein | 607869 |
Sensitivity | Light and moisture sensitive |
Solubility Information | Soluble in dimethyl sulfoxide. |
Formula Weight | 135.13 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 7-Deazahypoxanthine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'