missing translation for 'onlineSavingsMsg'
Läs mer

7-Deazapurine, TRC

Produktkod. 30361241
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361241 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. View return policy
Product Code. 30361241 Supplier Toronto Research Chemicals Supplier No. TRCD201950100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 7-Deazapurine
CAS 18549-65-0
Molekylformel C6 H5 N3
Kvantitet 100 mg
InChI formel InChI=1S/C6H5N3/c1-2-8-6-5(1)3-7-4-9-6/h1-4H,(H,7,8,9)
LEDER c1cc2c[nH]cnc2n1
IUPAC-namn 3H-pyrrolo[2,3-d]pyrimidine
Molekylvikt (g/mol) 119.12
Formel vikt 119.0483
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.