Läs mer
7-Dehydro Desmosterol, TRC
Produktkod. 30358119
Change view
Kvantitet:
0.5 mg
Produktkod. Kvantitet
30358119 0.5 mg
1 options
Produktkod. 30358119 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2295550.5MG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-Dehydro Desmosterol
CAS 1715-86-2
Molekylformel C27 H42 O
Kvantitet 0.5 mg
Synonym (3beta)-Cholesta-5,7,24-trien-3-ol, Cholesta-5,7,24-trien-3beta-ol, 7-Dehydrodesmosterol, Cholesta-5,7,24(25)-trien-3beta-ol, Cholesta-5,7,24-triene-3beta-ol
InChI formel InChI=1S/C27H42O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,9-10,19,21,23-25,28H,6,8,11-17H2,1-5H3/t19-,21+,23-,24+,25+,26+,27-/m1/s1
LEDER C[C@H](CCC=C(C)C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn (3S,9S,10R,13R,14R,17R)-10,13-dimethyl-17-[(2R)-6-methylhept-5-en-2-yl]-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvikt (g/mol) 382.622
Formel vikt 382.324