Läs mer
7-Descarboxamido, 7-Cyano (R)-Silodosin O-Benzoate Oxalate, TRC
Produktkod. 30361136
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361136 250 mg
30353465 25 mg
30353983 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30361136 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC824425250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 885340-12-5
Molekylformel C32H34F3N3O4 . C2H2O4
Molekylvikt (g/mol) 671.7
Synonym 1-[3-(Benzoyloxy)propyl]-2,3-dihydro-5-[(2R)-2-[[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl]amino]propyl]-1H-indole-7-carbonitrile ethanedioate (1:1), (R)-3-(7-Cyano-5-(2-((2-(2-(2,2,2-trifluoroethoxy)phenoxy)ethyl)amino)propyl)indolin-1-yl)propyl benzoate Oxalate
IUPAC-namn 3-[7-cyano-5-[(2R)-2-[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethylamino]propyl]-2,3-dihydroindol-1-yl]propyl benzoate;oxalic acid
LEDER C[C@H](Cc1cc2CCN(CCCOC(=O)c3ccccc3)c2c(c1)C#N)NCCOc4ccccc4OCC(F)(F)F.OC(=O)C(=O)O
Specifikationer
Kemiskt namn eller material 7-Descarboxamido, 7-Cyano (R)-Silodosin O-Benzoate Oxalate
Kvantitet 250 mg
InChI formel InChI=1S/C32H34F3N3O4.C2H2O4/c1-23(37-13-17-40-28-10-5-6-11-29(28)42-22-32(33,34)35)18-24-19-26-12-15-38(30(26)27(20-24)21-36)14-7-16-41-31(39)25-8-3-2-4-9-25;3-1(4)2(5)6/h2-6,8-11,19-20,23,37H,7,12-18,22H2,1H3;(H,3,4)(H,5,6)/t23-;/m1./s1
Formel vikt 671.2454