Läs mer
7-Desmethyl-3-hydroxyagomelatine, TRC
Produktkod. 30351953
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30351953 25 mg -
30351133 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30351953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28215025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 166527-00-0
Molekylformel C14H15NO3
Molekylvikt (g/mol) 245.27
Synonym N-[2-(3,7-dihydroxy-1-naphthalenyl)ethyl]acetamide, Agomelatine 7-Desmethyl-3-Hydroxy Impurity
IUPAC-namn N-(2-(3,7-dihydroxynaphthalen-1-yl)ethyl)acetamide
LEDER OC1=CC2=C(CCNC(C)=O)C=C(O)C=C2C=C1
Specifikationer
Kemiskt namn eller material 7-Desmethyl-3-hydroxyagomelatine
Kvantitet 25 mg
InChI formel InChI=1S/C14H15NO3/c1-9(16)15-5-4-11-7-13(18)6-10-2-3-12(17)8-14(10)11/h2-3,6-8,17-18H,4-5H2,1H3,(H,15,16)
Formel vikt 245.11