Learn More
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin, TRC
Product Code. 30375713
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30375713 25 mg 25mg
1 options
Product Code. 30375713 Supplier Toronto Research Chemicals Supplier No. TRCD47377825MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin
CAS 76877-33-3
Molecular Formula C24H22N2O4
Quantity 25 mg
Synonym 1-[4-[7-(Diethylamino)-4-methyl-2-oxo-2H-1-benzopyran-3-yl]phenyl]-1H-pyrrole-2,5-dione, 7-(Diethylamino)-3-(4'-maleimidylphenyl)-4-methylcoumarin, D 346
InChI Formula InChI=1S/C24H22N2O4/c1-4-25(5-2)18-10-11-19-15(3)23(24(29)30-20(19)14-18)16-6-8-17(9-7-16)26-21(27)12-13-22(26)28/h6-14H,4-5H2,1-3H3
SMILES CCN(CC)c1ccc2C(=C(C(=O)Oc2c1)c3ccc(cc3)N4C(=O)C=CC4=O)C
Molecular Weight (g/mol) 402.44