Läs mer
7-Epi Clindamycin, TRC
Produktkod. 30375634
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30375634 5 mg
1 options
Produktkod. 30375634 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5824855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-Epiclindamycin
CAS 16684-06-3
Anteckningar om renhetsgrad HPLC
Molekylformel C18 H33 Cl N2 O5 S
Kvantitet 5 mg
Synonym D-erythro-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, D-erythro-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-, trans- α- (8CI), D-erythro-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, (2S-trans)-, 7-Epiclindamycin, Antibiotic U 21251F, Methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-D-erythro-α-D-galacto-octopyranoside
InChI formel InChI=1S/C18H33ClN2O5S/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(26-16)27-4/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25)/t9-,10-,11+,12-,13+,14-,15-,16-,18-/m1/s1
LEDER CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O
IUPAC-namn (2S,4R)-N-[(1S,2R)-2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide
Molekylvikt (g/mol) 424.98
Formel vikt 424.1799