Läs mer
7-Ethoxy-4-(trifluoromethyl)coumarin, TRC
Produktkod. 30138497
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138497 250 mg
30012117 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138497 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE893055250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115453-82-2
Molekylformel C12H9F3O3
Molekylvikt (g/mol) 258.19
Synonym 7-Ethoxy-4-(trifluoromethyl)-2H-1-benzopyran-2-one, 4-(Trifluoromethyl)-7-ethoxycoumarin;
IUPAC-namn 7-ethoxy-4-(trifluoromethyl)-2H-chromen-2-one
LEDER O=C1OC=2C=C(OCC)C=CC2C(=C1)C(F)(F)F
Specifikationer
Kemiskt namn eller material 7-Ethoxy-4-(trifluoromethyl)coumarin
Kvantitet 250 mg
InChI formel InChI=1S/C12H9F3O3/c1-2-17-7-3-4-8-9(12(13,14)15)6-11(16)18-10(8)5-7/h3-6H,2H2,1H3
Formel vikt 258.05