Läs mer
7-Ethyl-10-(4-amino-1-piperidino)carbonyloxycamptothecin, TRC
Produktkod. 30012275
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012275 10 mg
30007745 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE89915010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 185304-42-1
Molekylformel C28 H30 N4 O6
Molekylvikt (g/mol) 518.56
LEDER CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(OC(=O)N6CCC(N)CC6)cc15)[C@@](O)(CC)C(=O)OC4
Specifikationer
Kemiskt namn eller material 7-Ethyl-10-(4-amino-1-piperidino)carbonyloxycamptothecin
Kvantitet 10 mg
InChI formel InChI=1S/C28H30N4O6/c1-3-17-18-11-16(38-27(35)31-9-7-15(29)8-10-31)5-6-22(18)30-24-19(17)13-32-23(24)12-21-20(25(32)33)14-37-26(34)28(21,36)4-2/h5-6,11-12,15,36H,3-4,7-10,13-14,29H2,1-2H3/t28-/m0/s1
Formel vikt 518.2165