missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorobenzo-2-oxa-1,3-diazole-4-sulfonic acid ammonium salt, MP Biomedicals™

Catalog Number p-4774482
Click to view available options
Quantity:
5 mg
10 mg
25 mg
This item is not returnable. View return policy
84806-27-9
C6H6FN3O4S
235.189
MFCD02682109
JXLHNMVSKXFWAO-UHFFFAOYSA-N
sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
2737780
azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]
This item is not returnable. View return policy
84806-27-9
C6H6FN3O4S
235.189
MFCD02682109
JXLHNMVSKXFWAO-UHFFFAOYSA-N
sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
2737780
azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]

Sensitive and specific fluorescent labeling reagent for low molecular weight compounds and for macromolecules containing thiol groups

Quantity 5 mg
Formula Weight 235.2
Physical Form Powder
Chemical Name or Material 7-Fluorobenzo-2-oxa-1, 3-diazole-4-sulfonic acid ammonium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'