Läs mer
7-[(Hydroxy-2-thienyl-3-thienylacetyl)oxy]-9,9-dimethyl-3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide, TRC
Produktkod. 30134374
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134374 50 mg
30009747 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134374 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH96202550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136431-94-2
Molekylformel C19H22BrNO4S2
Molekylvikt (g/mol) 472.42
Synonym Iso Tiotropium Bromide;
LEDER O=C(C(C1=CC=CS1)(O)C2=CSC=C2)O[C@H]3C[C+]4C(O5)C5[C@@H](C3)[N]4(C)C.[Br-]
Specifikationer
Kemiskt namn eller material 7-[(Hydroxy-2-thienyl-3-thienylacetyl)oxy]-9,9-dimethyl-3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide