Läs mer
7-Hydroxy Chlorpromazine Hydrochloride, TRC
Produktkod. 30012633
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30012633 25 mg -
30009762 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012633 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH82506525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51938-11-5
Molekylformel C17 H19 Cl N2 O S . HCl
Molekylvikt (g/mol) 371.33
Synonym 8-Chloro-10-[3-(dimethylamino)propyl]-10H-phenothiazin-3-ol Hydrochloride, 8-Chloro-10-[3-(dimethylamino)propyl]phenothiazin-3-ol Hydrochloride, 7-Hydroxy-2-chloropromazine, 7-Hydroxychlorpromazine Hydrochloride, NSC 131053 Hydrochloride, Chlorpromazine 7-Hydroxy Impurity
IUPAC-namn 8-chloro-10-[3-(dimethylamino)propyl]phenothiazin-3-ol;hydrochloride
LEDER ClC(C=C1)=CC2=C1SC(C=C(O)C=C3)=C3N2CCCN(C)C.Cl
Specifikationer
Kemiskt namn eller material 7-Hydroxy Chlorpromazine Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C17H19ClN2OS.ClH/c1-19(2)8-3-9-20-14-6-5-13(21)11-17(14)22-16-7-4-12(18)10-15(16)20;/h4-7,10-11,21H,3,8-9H2,1-2H3;1H
Formel vikt 370.0673
Procent renhet >95%