Läs mer
7-Ketoabiraterone Acetate, TRC
Produktkod. 30134093
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134093 100 mg
30007845 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134093 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK170980100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2410075-48-6
Molekylformel C26H31NO3
Molekylvikt (g/mol) 405.53
Synonym (3S,8R,9S,10R,13S,14S)-10,13-Dimethyl-7-oxo-17-(pyridin-3-yl)-2,3,4,7,8,9,10,11,12,13,14,15-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl Acetate
IUPAC-namn [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-7-oxo-17-(3-pyridyl)-1,2,3,4,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-3-yl] acetate
LEDER CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4c5cccnc5)[C@@H]3C(=O)C=C2C1
Specifikationer
Kemiskt namn eller material 7-Ketoabiraterone Acetate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C26H31NO3/c1-16(28)30-19-8-10-25(2)18(13-19)14-23(29)24-21-7-6-20(17-5-4-12-27-15-17)26(21,3)11-9-22(24)25/h4-6,12,14-15,19,21-22,24H,7-11,13H2,1-3H3/t19-,21-,22-,24-,25-,26+/m0/s1
Formel vikt 405.2304
Procent renhet >95%