Läs mer
7-Methoxy-1-naphthaleneacetamide, TRC
Produktkod. 30472062
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472062 2.5 g
30480539 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30472062 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2647952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 138113-07-2
Molekylformel C13 H13 N O2
Molekylvikt (g/mol) 215.25
Synonym Agomelatine Impurity B
IUPAC-namn 2-(7-methoxynaphthalen-1-yl)acetamide
LEDER COc1ccc2cccc(CC(=O)N)c2c1
Specifikationer
Kemiskt namn eller material 2-(7-methoxynaphthalen-1-yl)acetamide
Kvantitet 2.5 g
InChI formel InChI=1S/C13H13NO2/c1-16-11-6-5-9-3-2-4-10(7-13(14)15)12(9)8-11/h2-6,8H,7H2,1H3,(H2,14,15)
Formel vikt 215.0946