Learn More
Alfa Aesar™ 7-Methyl-4-indanol, 97%
Catalog Number p-7025034
Click to view available options
Quantity:
2.5 g
10 g
16400-13-8
C10H12O
148.21
MFCD00173675
XHMLXYGITDAGDN-UHFFFAOYSA-N
7-methylindan-4-ol, 4-hydroxy-7-methylindane, 1h-inden-4-ol,2,3-dihydro-7-methyl, 7-methyl-4-indanol, acmc-20amgr, 7-methyl-indan-4-ol
85398
7-methyl-2,3-dihydro-1H-inden-4-ol
CC1=CC=C(O)C2=C1CCC2
16400-13-8
C10H12O
148.21
MFCD00173675
XHMLXYGITDAGDN-UHFFFAOYSA-N
7-methylindan-4-ol, 4-hydroxy-7-methylindane, 1h-inden-4-ol,2,3-dihydro-7-methyl, 7-methyl-4-indanol, acmc-20amgr, 7-methyl-indan-4-ol
85398
7-methyl-2,3-dihydro-1H-inden-4-ol
CC1=CC=C(O)C2=C1CCC2
Applications
7-Methyl-4-indanol is used as an intermediate in synthetic chemistry.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 80°C to 82°C
Quantity 2.5 g
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material 7-Methyl-4-indanol