Läs mer
7-Methyl-7H-purin-6-amine, TRC
Produktkod. 18050070
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18050070 50 mg
18051650 5 mg
18052950 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18050070 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31460550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 935-69-3
Molekylformel C6 H7 N5
Molekylvikt (g/mol) 149.153
Synonym 7-Methyl-adenine, 6-Amino-7-methylpurine, 7-Methyladenine, N7-Methyladenine, NSC 7857
IUPAC-namn 7-methylpurin-6-amine
LEDER Cn1cnc2ncnc(N)c12
Specifikationer
Kemiskt namn eller material 7-Methyl-7H-purin-6-amine
Kvantitet 50 mg
InChI formel InChI=1S/C6H7N5/c1-11-3-10-6-4(11)5(7)8-2-9-6/h2-3H,1H3,(H2,7,8,9)
Formel vikt 149.0701