Läs mer
7-Methylguanine, TRC
Produktkod. 30006764
Change view
Click to view available options
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30006764 250 mg
30007227 1 g
17910905 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006764 Supplier Toronto Research Chemicals Supplier No. TRCM311790250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 578-76-7
Molekylformel C6 H7 N5 O
Molekylvikt (g/mol) 165.15
Synonym 2-Amino-1,7-dihydro-7-methyl-6H-purin-6-one, 2-Amino-7-methylhypoxanthine, N7-Methylguanine, NSC 193444, NSC 19647
IUPAC-namn 2-amino-7-methyl-1H-purin-6-one
LEDER Cn1cnc2N=C(N)NC(=O)c12
Specifications
Kemiskt namn eller material 7-Methylguanine
Kvantitet 250 mg
InChI formel InChI=1S/C6H7N5O/c1-11-2-8-4-3(11)5(12)10-6(7)9-4/h2H,1H3,(H3,7,9,10,12)
Formel vikt 165.0651