missing translation for 'onlineSavingsMsg'
Läs mer

7-Methyltryptophol, TRC

Produktkod. 30480395
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480395 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30480395 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3318005G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 7-Methyltryptophol
CAS 39232-85-4
Molekylformel C11H13NO
Kvantitet 5 g
Synonym 2-(7-Methyl-1H-indol-3-yl)ethanol, 7-Methyltryptophol
InChI formel InChI=1S/C11H13NO/c1-8-3-2-4-10-9(5-6-13)7-12-11(8)10/h2-4,7,12-13H,5-6H2,1H3
LEDER Cc1cccc2c(CCO)c[nH]c12
IUPAC-namn 2-(7-methyl-1H-indol-3-yl)ethanol
Molekylvikt (g/mol) 175.23
Formel vikt 175.0997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.