missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Nitro-1H-indazole, 98%, Thermo Scientific Chemicals
510.45 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Beschreibung
7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Spezifikation
Melting Point | 184°C to 189°C |
Quantity | 1 g |
Beilstein | 6809 |
Solubility Information | Insoluble in water. |
Formula Weight | 163.14 |
Percent Purity | 98% |
Chemical Name or Material | 7-Nitro-1H-indazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'