missing translation for 'onlineSavingsMsg'
Läs mer

7-Nitrosancycline Monosulfate, TRC

Produktkod. 30471101
Change view
Click to view available options
Kvantitet:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30471101 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471101 Supplier Toronto Research Chemicals Supplier No. TRCN5210002MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 7-Nitrosancycline Monosulfate
CAS 5679-01-6
Molekylformel C21H23N3O13S
Kvantitet 2 mg
Synonym 7-Nitro-6-demethyl-6-deoxytetracycline Sulfate, [4S-(4alpha,4aalpha,5aalpha,12aalpha)]-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-7-nitro-1,11-dioxo-naphthacenecarboxamide Sulfate
InChI formel InChI=1S/C21H21N3O9.H2O4S/c1-23(2)15-9-6-7-5-8-10(24(32)33)3-4-11(25)13(8)16(26)12(7)18(28)21(9,31)19(29)14(17(15)27)20(22)30;1-5(2,3)4/h3-4,7,9,15,25,27-28,31H,5-6H2,1-2H3,(H2,22,30);(H2,1,2,3,4)/t7-,9-,15-,21-;/m0./s1
LEDER CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4c(C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)c(O)ccc4[N+](=O)[O-].OS(=O)(=O)O
IUPAC-namn (4S,4aS,5aR,12aS)-4-(dimethylamino)-3,10,12,12a-tetrahydroxy-7-nitro-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;sulfuric acid
Molekylvikt (g/mol) 557.48
Formel vikt 557.0952
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.