Läs mer
7-O-Acetyl Ursodeoxycholic Acid Methyl Ester, TRC
Produktkod. 30316547
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30316547 50 mg
1 options
Produktkod. 30316547 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18976050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-O-Acetyl Ursodeoxycholic Acid Methyl Ester
CAS 75672-24-1
Molekylformel C27H44O5
Kvantitet 50 mg
Synonym (3alpha,5beta,7beta)-7-(Acetyloxy)-3-hydroxy-cholan-24-oic Acid Methyl Ester
InChI formel InChI=1S/C27H44O5/c1-16(6-9-24(30)31-5)20-7-8-21-25-22(11-13-27(20,21)4)26(3)12-10-19(29)14-18(26)15-23(25)32-17(2)28/h16,18-23,25,29H,6-15H2,1-5H3/t16-,18+,19-,20-,21+,22+,23+,25+,26+,27-/m1/s1
LEDER COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)C
IUPAC-namn methyl (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-7-acetyloxy-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molekylvikt (g/mol) 448.64
Formel vikt 448.3189