missing translation for 'onlineSavingsMsg'
Läs mer

7-O-Demethyl Rapamycin (~80%), TRC

Produktkod. 30354768
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354768 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30354768 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2312402.5MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 7-O-Demethyl Rapamycin
CAS 151519-50-5
Molekylformel C50H77NO13
Kvantitet 2.5 mg
Synonym 7-O-Demethyl Cypher, 7-O-Demethyl Rapammune, 7-O-Demethyl Sirolimus
InChI formel InChI=1S/C50H77NO13/c1-29-15-11-10-12-16-30(2)40(53)27-37-20-18-35(7)50(60,64-37)47(57)48(58)51-22-14-13-17-38(51)49(59)63-42(32(4)25-36-19-21-39(52)43(26-36)61-8)28-41(54)31(3)24-34(6)45(56)46(62-9)44(55)33(5)23-29/h10-12,15-16,24,29,31-33,35-40,42-43,45-46,52-53,56,60H,13-14,17-23,25-28H2,1-9H3/b12-10+,15-11+,30-16+,34-24+/t29-,31?,32-,33-,35-,36+,37+,38+,39-,40+,42+,43-,45-,46+,50-/m1/s1
LEDER CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(/C)\[C@@H](O)C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)CC[C@H]1O
Molekylvikt (g/mol) 900.15
Formel vikt 899.5395
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.