Läs mer
7-O-Methyl Quercetin, TRC
Produktkod. 17983885
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17983885 50 mg
17944145 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17983885 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32655550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90-19-7
Molekylformel C16 H12 O7
Molekylvikt (g/mol) 316.26
Synonym 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-3,5-dihydroxy-7-methoxy-, Flavone, 3,3',4',5-tetrahydroxy-7-methoxy- (7CI,8CI), Rhamnetin (6CI), 2-(3,4-Dihydroxyphenyl)-3,5-dihydroxy-7-methoxy-4H-1-benzopyran-4-one, 2-(3,4-Dihydroxyphenyl)-3,5-dihydroxy-7-methoxychromen-4-one, 3,3',4',5-Tetrahydroxy-7-methoxyflavone, 3,5,3',4'-Tetrahydroxy-7-methoxyflavone, 7-Methoxyquercetin, 7-Methylquercetin, 7-O-Methylquercetin, C.I. 75690, LY 805921, NSC 19802, Quercetin 7-methyl ether, β-Rhamnocitrin
IUPAC-namn 2-(3,4-dihydroxyphenyl)-3,5-dihydroxy-7-methoxychromen-4-one
LEDER COc1cc(O)c2C(=O)C(=C(Oc2c1)c3ccc(O)c(O)c3)O
Specifikationer
Kemiskt namn eller material Rhamnetin
Kvantitet 50 mg
InChI formel InChI=1S/C16H12O7/c1-22-8-5-11(19)13-12(6-8)23-16(15(21)14(13)20)7-2-3-9(17)10(18)4-7/h2-6,17-19,21H,1H3
Formel vikt 316.0583