Läs mer
7-Oxooctadecanoic Acid Methyl Ester, TRC
Produktkod. 30423949
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423949 50 mg
30424316 250 mg
30418527 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30423949 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO87016050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2380-22-5
Molekylformel C19H36O3
Molekylvikt (g/mol) 312.49
Synonym Methyl 7-Ketooctadecanoate, Methyl 7-Oxooctadecanoate
IUPAC-namn methyl 7-oxooctadecanoate
LEDER CCCCCCCCCCCC(=O)CCCCCC(=O)OC
Specifikationer
Kemiskt namn eller material 7-Oxooctadecanoic Acid Methyl Ester
Kvantitet 50 mg
InChI formel InChI=1S/C19H36O3/c1-3-4-5-6-7-8-9-10-12-15-18(20)16-13-11-14-17-19(21)22-2/h3-17H2,1-2H3
Formel vikt 312.2664