Läs mer
7-Phenylacetamide-3-chloromethyl-3-cephem-4-carboxylic Acid Diphenylmethyl Ester, TRC
Produktkod. 30428206
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428206 5 g
30419682 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30428206 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3191005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-Phenylacetamide-3-chloromethyl-3-cephem-4-carboxylic Acid
CAS 64308-63-0
Molekylformel C29 H25 Cl N2 O4 S
Kvantitet 5 g
InChI formel InChI=1S/C29H25ClN2O4S/c30-17-22-18-37-28-24(31-23(33)16-19-10-4-1-5-11-19)27(34)32(28)25(22)29(35)36-26(20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-15,24,26,28H,16-18H2,(H,31,33)/t24-,28-/m1/s1
LEDER ClCC1=C(N2[C@H](SC1)[C@H](NC(=O)Cc3ccccc3)C2=O)C(=O)OC(c4ccccc4)c5ccccc5
IUPAC-namn benzhydryl (6R,7R)-3-(chloromethyl)-8-oxo-7-[(2-phenylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molekylvikt (g/mol) 533.04
Formel vikt 532.1224