Läs mer
7-Sulfo-ursodeoxycholic Acid Disodium Salt, TRC
Produktkod. 17981435
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981435 1 mg
17943145 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17981435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS7892551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 191286-16-5
Molekylformel C24 H40 O7 S . 2H Na
Molekylvikt (g/mol) 520.63
Synonym (3α,5β,7β)-7-Hydroxy-3-(sulfooxy)-cholan-24-oic Acid Disodium Salt
IUPAC-namn (4R)-4-[(3R,5R,7S,8R,9S,10S,13R,14S,17R)-7-hydroxy-10,13-dimethyl-3-sulfooxy-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid;sodium hydride
LEDER [Na].[Na].C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material 7-Sulfo-ursodeoxycholic Acid Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C24H40O7S.2Na.2H/c1-14(4-7-21(26)27)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(31-32(28,29)30)12-15(23)13-20(22)25;;;;/h14-20,22,25H,4-13H2,1-3H3,(H,26,27)(H,28,29,30);;;;/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-;;;;/m1..../s1
Formel vikt 520.2447
Procent renhet >95%