missing translation for 'onlineSavingsMsg'
Läs mer

7-tert-Butyl-6-chloro-3-(2,5-difluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine, TRC

Produktkod. 30335382
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30335382 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30335382 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69061025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 7-tert-Butyl-6-chloro-3-(2,5-difluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine
CAS 286456-54-0
Molekylformel C15 H13 Cl F2 N4
Kvantitet 25 mg
Synonym 1,2,4-Triazolo[4,3-b]pyridazine, 6-chloro-3-(2,5-difluorophenyl)-7-(1,1-dimethylethyl)-, 6-Chloro-3-(2,5-difluorophenyl)-7-(1,1-dimethylethyl)-1,2,4-triazolo[4,3-b]pyridazine, 7-tert-Butyl-6-chloro-3-(2,5-difluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine
InChI formel InChI=1S/C15H13ClF2N4/c1-15(2,3)10-7-12-19-20-14(22(12)21-13(10)16)9-6-8(17)4-5-11(9)18/h4-7H,1-3H3
LEDER CC(C)(C)c1cc2nnc(c3cc(F)ccc3F)n2nc1Cl
IUPAC-namn 7-tert-butyl-6-chloro-3-(2,5-difluorophenyl)-[1,2,4]triazolo[4,3-b]pyridazine
Molekylvikt (g/mol) 322.74
Formel vikt 322.0797
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.