Learn More
7-(Trifluoromethyl)1H-indole-2,3-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 7-(Trifluoromethyl)1H-indole-2,3-dione |
| CAS | 391-12-8 |
| Molecular Formula | C9 H4 F3 N O2 |
| Quantity | 25 g |
| InChI Formula | InChI=1S/C9H4F3NO2/c10-9(11,12)5-3-1-2-4-6(5)13-8(15)7(4)14/h1-3H,(H,13,14,15) |
| SMILES | FC(F)(F)c1cccc2C(=O)C(=O)Nc12 |
| IUPAC Name | 7-(trifluoromethyl)-1H-indole-2,3-dione |
| Molecular Weight (g/mol) | 215.13 |
| Formula Weight | 215.0194 |